accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3366-93-6

CH2NOHNO

CAS RN 3366-93-6

IUPAC Name: 1-(prop-2-en-1-yl)imidazolidine-2,4-dione
Synonyms: 1-(prop-2-en-1-yl)imidazolidine-2,4-dione
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: KPDTTZWHFZUVCL-UHFFFAOYSA-N
SMILES: C=CCN1CC(=O)NC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results