accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OCH3OOH3C

CAS RN 33697-53-9

IUPAC Name: ethyl 2-ethyl-2-methyl-3-oxobutanoate
Synonyms: ethyl 2-ethyl-2-methyl-3-oxobutanoate
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: NWMTWESXONQJPU-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)(CC)C(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results