Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Ethyl-2-methylacetoacetate 93.0+%, TCI America™
SDP

Catalog No. E04375ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04375ML 5 mL
E043725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04375ML Supplier TCI America Supplier No. E04375ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33697-53-9
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
MDL Number MFCD00191433
InChI Key NWMTWESXONQJPU-UHFFFAOYSA-N
Synonym 2-Ethyl-2-methylacetoacetic Acid Ethyl Ester
PubChem CID 582452
IUPAC Name ethyl 2-ethyl-2-methyl-3-oxobutanoate
SMILES CCC(C)(C(=O)C)C(=O)OCC

Specifications

Color Yellow
Boiling Point 199°C
Quantity 5 mL
Formula Weight 172.22
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Ethyl-2-methylacetoacetate
EINECSNumber (2)-1505
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.