CAS RN 33795-85-6
IUPAC Name:
4,5,6,7-tetrafluoro-2-methyl-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms:
4,5,6,7-tetrafluoro-2-methylisoindole-1,3-dione
Molecular Weight (g/mol):
233.12
Molecular Formula:
C9H3F4NO2
InChi Key:
AULXVJOTNZUFIY-UHFFFAOYSA-N
SMILES:
CN1C(=O)C2=C(C1=O)C(F)=C(F)C(F)=C2F
1
–
1
of
1
results
1
3,4,5,6-Tetrafluoro-N-methylphthalimide 95.0+%, TCI America™
CAS: 33795-85-6 Molecular Formula: C9H3F4NO2 Molecular Weight (g/mol): 233.12 MDL Number: MFCD01631699 InChI Key: AULXVJOTNZUFIY-UHFFFAOYSA-N PubChem CID: 469561 IUPAC Name: 4,5,6,7-tetrafluoro-2-methyl-2,3-dihydro-1H-isoindole-1,3-dione SMILES: CN1C(=O)C2=C(C1=O)C(F)=C(F)C(F)=C2F