CAS RN 3386-97-8
IUPAC Name:
4-isothiocyanatobut-1-ene
Synonyms:
1-butene, 4-isothiocyanato-
Molecular Weight (g/mol):
113.18
Molecular Formula:
C5H7NS
InChi Key:
SKIHGKNFJKJXPX-UHFFFAOYSA-N
SMILES:
C=CCCN=C=S
1
–
1
of
1
results
1
3-Buten-1-yl Isothiocyanate 96.0+%, TCI America™
CAS: 3386-97-8 Molecular Formula: C5H7NS Molecular Weight (g/mol): 113.178 MDL Number: MFCD00191660 InChI Key: SKIHGKNFJKJXPX-UHFFFAOYSA-N Synonym: Isothiocyanic Acid 3-Buten-1-yl Ester PubChem CID: 76922 IUPAC Name: 4-isothiocyanatobut-1-ene SMILES: C=CCCN=C=S