accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiBONH3CONCH3ONH3CONCH3

CAS RN 338949-42-1

IUPAC Name: lithium(1+) tetrakis[(2-methylquinolin-8-yl)oxy]boranuide
Synonyms: lithium(1+) tetrakis[(2-methylquinolin-8-yl)oxy]boranuide
Molecular Weight (g/mol): 650.47
Molecular Formula: C40H32BLiN4O4
InChi Key: AVPRLXSPVRRDIE-UHFFFAOYSA-N
SMILES: [Li+].CC1=NC2=C(O[B-](OC3=C4N=C(C)C=CC4=CC=C3)(OC3=C4N=C(C)C=CC4=CC=C3)OC3=C4N=C(C)C=CC4=CC=C3)C=CC=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results