accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33918-15-9

IUPAC Name: 3-[(propan-2-yl)amino]propan-1-ol
Synonyms: 3-(isopropylamino)propan-1-ol
Molecular Weight (g/mol): 117.19
Molecular Formula: C6H15NO
InChi Key: GZCPWFOPXIDRDP-UHFFFAOYSA-N
SMILES: CC(C)NCCCO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results