Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Isopropylamino)propanol 98.0+%, TCI America™
SDP

Catalog No. I07555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07555G 5 g
I075525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07555G Supplier TCI America Supplier No. I07555G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33918-15-9
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.192
MDL Number MFCD00483086
InChI Key GZCPWFOPXIDRDP-UHFFFAOYSA-N
PubChem CID 2063457
IUPAC Name 3-(propan-2-ylamino)propan-1-ol
SMILES CC(C)NCCCO

Specifications

Color White
Boiling Point 94°C
Quantity 5 g
Formula Weight 117.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-(Isopropylamino)propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.