CAS RN 3392-11-8
IUPAC Name:
2,5-dioxopyrrolidin-1-yl 2-{[(tert-butoxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoate
Synonyms:
2,5-dioxopyrrolidin-1-yl 2-[(tert-butoxycarbonyl)amino]-3-(1H-indol-3-yl)propanoate
Molecular Weight (g/mol):
401.42
Molecular Formula:
C20H23N3O6
InChi Key:
CPJXMXQYRHNIFU-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CC1=CNC2=CC=CC=C12)C(=O)ON1C(=O)CCC1=O
1
–
1
of
1
results
1
Nalpha-(tert-Butoxycarbonyl)-L-tryptophan N-Succinimidyl Ester, TCI America™
CAS: 3392-11-8 Molecular Formula: C20H23N3O6 Molecular Weight (g/mol): 401.42 MDL Number: MFCD00022756 InChI Key: CPJXMXQYRHNIFU-UHFFFAOYNA-N Synonym: Nalpha-Boc-L-tryptophan N-Succinimidyl Ester, Boc-Trp-OSu PubChem CID: 98767 IUPAC Name: (2,5-dioxopyrrolidin-1-yl) (2S)-3-(1H-indol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate SMILES: CC(C)(C)OC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)ON3C(=O)CCC3=O