accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 339531-50-9

IUPAC Name: (4S)-3-[(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
Synonyms: (4S)-3-[(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-methylpentanoyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
Molecular Weight (g/mol): 480.56
Molecular Formula: C27H32N2O6
InChi Key: BUYLKVIICSUHDX-VVWGFWRPNA-N
SMILES: CC(C)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)N1[C@@H](COC1(C)C)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)