accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOSOO

CAS RN 3406-02-8

IUPAC Name: (benzenesulfonyl)methanesulfonylbenzene
Synonyms: (benzenesulfonyl)methanesulfonylbenzene
Molecular Weight (g/mol): 296.36
Molecular Formula: C13H12O4S2
InChi Key: QCHNSJNRFSOCLJ-UHFFFAOYSA-N
SMILES: O=S(=O)(CS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results