Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(phenylsulfonyl)methane 98.0+%, TCI America™
SDP

Catalog No. B21675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21675G Supplier TCI America Supplier No. B21675G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3406-02-8
Molecular Formula C13H12O4S2
Molecular Weight (g/mol) 296.36
MDL Number MFCD00007553
InChI Key QCHNSJNRFSOCLJ-UHFFFAOYSA-N
Synonym bis phenylsulfonyl methane, methylenebis sulphonyl bisbenzene, benzene, 1,1'-methylenebis sulfonyl bis, bis benzenesulfonyl methane, benzenesulfonyl methanesulfonylbenzene, phenylsulfonyl methyl sulfonyl benzene, bisphenylsulfonylmethane, maybridge3_003227, acmc-1cmc8, bis phenylsulphonyl methane
PubChem CID 76948
IUPAC Name benzenesulfonylmethylsulfonylbenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)CS(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 5 g
Formula Weight 296.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(phenylsulfonyl)methane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.