CAS RN 34151-49-0
IUPAC Name:
6,13-bis(pyridin-4-yl)-6,13-diazatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone
Synonyms:
6,13-bis(pyridin-4-yl)-6,13-diazatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone
Molecular Weight (g/mol):
420.38
Molecular Formula:
C24H12N4O4
InChi Key:
IBRPEOCBRYYINT-UHFFFAOYSA-N
SMILES:
O=C1N(C2=CC=NC=C2)C(=O)C2=CC=C3C(=O)N(C4=CC=NC=C4)C(=O)C4=CC=C1C2=C34
1
–
1
of
1
results
1
N,N'-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide 98.0+%, TCI America™
CAS: 34151-49-0 Molecular Formula: C24H12N4O4 Molecular Weight (g/mol): 420.384 InChI Key: IBRPEOCBRYYINT-UHFFFAOYSA-N PubChem CID: 1381898 SMILES: C1=CC2=C3C(=CC=C4C3=C1C(=O)N(C4=O)C5=CC=NC=C5)C(=O)N(C2=O)C6=CC=NC=C6