CAS RN 34369-07-8
Thermo Scientific Chemicals Adenosine-5'-triphosphate disodium salt hydrate, 99%, water <10%
CAS: 34369-07-8 Molecular Formula: C10H14N5Na2O13P3 Molecular Weight (g/mol): 551.15 MDL Number: MFCD00150755 InChI Key: TTWYZDPBDWHJOR-WCYUCLFNNA-L Synonym: atp disodium salt hydrate,atp disodium salt,atp disodium hydrate,adenosine 5'-triphosphate disodium salt hydrate,atp disodium salt trihydrate,56-65-5 non-sodium,987-65-5 anhydrous,5 inverted exclamation marka-atp-na2,adenosine 5'-triphosphate disodium sal PubChem CID: 131664345 IUPAC Name: disodium;[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl] phosphate;hydrate SMILES: [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
Adenosine 5'-Triphosphate Disodium Salt Hydrate 98.0+%, TCI America™
CAS: 34369-07-8 Molecular Formula: C10H14N5Na2O13P3 Molecular Weight (g/mol): 551.15 MDL Number: MFCD00150755 InChI Key: TTWYZDPBDWHJOR-WCYUCLFNNA-L Synonym: atp disodium salt hydrate,atp disodium salt,atp disodium hydrate,adenosine 5'-triphosphate disodium salt hydrate,atp disodium salt trihydrate,56-65-5 non-sodium,987-65-5 anhydrous,5 inverted exclamation marka-atp-na2,adenosine 5'-triphosphate disodium sal PubChem CID: 131664345 IUPAC Name: disodium phosphono ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate SMILES: [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
Thermo Scientific Chemicals Adenosine-5'-triphosphate disodium salt hydrate, ultrapure, 98%
CAS: 34369-07-8 Molecular Formula: C10H14N5Na2O13P3 Molecular Weight (g/mol): 551.15 MDL Number: MFCD00150755 InChI Key: TTWYZDPBDWHJOR-WCYUCLFNNA-L Synonym: atp disodium salt hydrate,atp disodium salt,atp disodium hydrate,adenosine 5'-triphosphate disodium salt hydrate,atp disodium salt trihydrate,56-65-5 non-sodium,987-65-5 anhydrous,5 inverted exclamation marka-atp-na2,adenosine 5'-triphosphate disodium sal PubChem CID: 131664345 IUPAC Name: disodium;[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl] phosphate;hydrate SMILES: [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
Thermo Scientific Chemicals Adenosine-5'-triphosphate disodium salt hydrate, 98%
CAS: 34369-07-8 Molecular Formula: C10H14N5Na2O13P3 Molecular Weight (g/mol): 551.15 MDL Number: MFCD00150755 InChI Key: TTWYZDPBDWHJOR-WCYUCLFNNA-L Synonym: atp disodium salt hydrate,atp disodium salt,atp disodium hydrate,adenosine 5'-triphosphate disodium salt hydrate,atp disodium salt trihydrate,56-65-5 non-sodium,987-65-5 anhydrous,5 inverted exclamation marka-atp-na2,adenosine 5'-triphosphate disodium sal PubChem CID: 131664345 IUPAC Name: disodium;[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl] phosphate;hydrate SMILES: [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
Adenosine 5'-triphosphate, disodium salt hydrate, 98%
CAS: 34369-07-8 Molecular Formula: C10H14N5Na2O13P3 Molecular Weight (g/mol): 551.15 MDL Number: MFCD00150755 InChI Key: TTWYZDPBDWHJOR-WCYUCLFNNA-L PubChem CID: 131664345 IUPAC Name: disodium phosphono ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate SMILES: [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
Sigma Aldrich Adenosine 5'-Triphosphate Disodium Salt Hydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich N-Acetylglycine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Adenosine 5'-triphosphate disodium salt hydrate Thermo Scientific Chemicals
CAS: 34369-07-8 Molecular Formula: C10H20N5Na2O16P3 Molecular Weight (g/mol): 605.19 MDL Number: MFCD00150754 InChI Key: MWEQTWJABOLLOS-MVHBFMHHNA-L Synonym: adenosine 5 '-triphosphate disodium salt trihydrate,adenosine 5 '-triphosphate disodium salt,adenosine 5 '-triphosphate disodium salt trihydrate, disodium salt PubChem CID: 134129625 IUPAC Name: ({[(2 R,3 S,4 R,5 R)-5-(6-amino-9 H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(phosphonatooxy)phosphinate SMILES: O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1