Learn More
Adenosine 5'-triphosphate disodium salt hydrate Thermo Scientific Chemicals
Shop All Thermo Scientific ProductsSpecifications
Specifications
| Physical Form | Crystalline Powder |
| Quantity | 5 g |
| Formula Weight | 551.14 (Anhydrous) |
| Packaging | Plastic bottle |
| Chemical Name or Material | Adenosine-5 '-Triphosphate Disodium Salt Trihydrate |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.