accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClBrCl

CAS RN 343781-45-3

IUPAC Name: 4-bromo-3,5-dichloropyridine
Synonyms: 4-bromo-3,5-dichloropyridine
Molecular Weight (g/mol): 226.88
Molecular Formula: C5H2BrCl2N
InChi Key: LOKLQBZBVPYAQO-UHFFFAOYSA-N
SMILES: ClC1=CN=CC(Cl)=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results