accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3440-99-1

IUPAC Name: 5-(3-bromophenyl)-2H-1,2,3,4-tetrazole
Synonyms: 5-(3-bromophenyl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 225.05
Molecular Formula: C7H5BrN4
InChi Key: UVKPUDRFBHSFJH-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results