CAS RN 34452-51-2
IUPAC Name:
potassium 2-sulfanylacetate
Synonyms:
potassium thioglycolate
Molecular Weight (g/mol):
130.2
Molecular Formula:
C2H3KO2S
InChi Key:
HYTYHTSMCRDHIM-UHFFFAOYSA-M
SMILES:
[K+].[O-]C(=O)CS
1
–
1
of
1
results
1
MP Biomedicals, Inc Potassium Thioglycollate, 40% Soln, MP Biomedicals
CAS: 34452-51-2 Molecular Formula: C2H3KO2S Molecular Weight (g/mol): 130.202 InChI Key: HYTYHTSMCRDHIM-UHFFFAOYSA-M Synonym: potassium thioglycolate,unii-xp5wzo5w6h,potassium 2-mercaptoacetate,acetic acid, mercapto-, monopotassium salt,xp5wzo5w6h,68-11-1 parent,potassium sulfanylacetate,potassium mercaptoacetate,kopcolate,potassium ion thioglycolate PubChem CID: 23688957 IUPAC Name: potassium;2-sulfanylacetate SMILES: C(C(=O)[O-])S.[K+]