accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34562-31-7

IUPAC Name: 3,5-diethyl-1-phenyl-2-propyl-1,2-dihydropyridine
Synonyms: 3,5-diethyl-1-phenyl-2-propyl-2H-pyridine
Molecular Weight (g/mol): 255.41
Molecular Formula: C18H25N
InChi Key: IAAASXBHFUJLHW-UHFFFAOYNA-N
SMILES: CCCC1N(C=C(CC)C=C1CC)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)