Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diethyl-1,2-dihydro-1-phenyl-2-n-propylpyridine, tech. 75%, Thermo Scientific™

Catalog No. AAH3253818
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3253818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3253818 Supplier Thermo Scientific Chemicals Supplier No. H3253818

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34562-31-7
Molecular Formula C18H25N
Molecular Weight (g/mol) 255.41
MDL Number MFCD00071789
InChI Key IAAASXBHFUJLHW-UHFFFAOYNA-N
Synonym 3,5-diethyl-1-phenyl-2-propyl-1,2-dihydropyridine, 3,5-diethyl-1,2-dihydro-1-phenyl-2-propylpyridine, pdhp, pyridine, 3,5-diethyl-1,2-dihydro-1-phenyl-2-propyl, 1-phenyl-3,5-diethyl-2-propyl-1,2-dihydropyridine, dsstox_cid_24923, dsstox_rid_80589, dsstox_gsid_44923, 3,5-diethyl-1,2-dihydro-1-phenyl-2-n-propylpyridine, 3,5-diethyl-1,2-dihydro-1-phenyl-2-propyl pyridine
PubChem CID 118703
IUPAC Name 3,5-diethyl-1-phenyl-2-propyl-2H-pyridine
SMILES CCCC1N(C=C(CC)C=C1CC)C1=CC=CC=C1

Specifications

Density 0.97
Boiling Point 125°C (0.5 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.5675
Quantity 50 g
Formula Weight 255.4
Percent Purity 75%
Grade Technical
Chemical Name or Material 3,5-Diethyl-1,2-dihydro-1-phenyl-2-n-propylpyridine
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 252-091-3
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.