accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OSSNHONHNN

CAS RN 345627-80-7

IUPAC Name: N-(5-{[(5-tert-butyl-1,3-oxazol-2-yl)methyl]sulfanyl}-1,3-thiazol-2-yl)piperidine-4-carboxamide
Synonyms: N-(5-{[(5-tert-butyl-1,3-oxazol-2-yl)methyl]sulfanyl}-1,3-thiazol-2-yl)piperidine-4-carboxamide
Molecular Weight (g/mol): 380.53
Molecular Formula: C17H24N4O2S2
InChi Key: OUSFTKFNBAZUKL-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CN=C(CSC2=CN=C(NC(=O)C3CCNCC3)S2)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results