accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONOCH3HNNH2OH(E/Z)

CAS RN 345893-91-6

IUPAC Name: 3-{1-[(2-aminophenyl)amino]ethylidene}-1-hydroxy-4-methyl-1,2,3,6-tetrahydropyridine-2,6-dione
Synonyms: 3-{1-[(2-aminophenyl)amino]ethylidene}-1-hydroxy-4-methylpyridine-2,6-dione
Molecular Weight (g/mol): 273.29
Molecular Formula: C14H15N3O3
InChi Key: IMUCOECFKIWBSW-UHFFFAOYSA-N
SMILES: CC(NC1=CC=CC=C1N)=C1C(C)=CC(=O)N(O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)