accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SSOOHH3CH3CCH3

CAS RN 345952-44-5

IUPAC Name: 6-[2-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)-1,3-dithiolan-2-yl]naphthalene-2-carboxylic acid
Synonyms: 6-[2-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)-1,3-dithiolan-2-yl]naphthalene-2-carboxylic acid
Molecular Weight (g/mol): 462.67
Molecular Formula: C28H30O2S2
InChi Key: DPVGXUNWCVEIGT-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=C1C=CC(=C2)C1(SCCS1)C1=CC=C2C=C(C=CC2=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results