Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MM 11253

Catalog No. 382250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-225-0 50 mg
38-2210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-225-0 Supplier Tocris Bioscience™ Supplier No. 3822/50
This item is not returnable. View return policy

RARγ-selective antagonist

Selective RARγ antagonist. Blocks the growth inhibitory ability of RARγ-selective agonists in squamous cell carcinoma (SCC)-25 cells.

Chemical Identifiers

CAS 345952-44-5
Molecular Formula C28H30O2S2

Specifications

Quantity 50 mg
Formula Weight 462.67
Purity >98%
Molecular Weight (g/mol) 462.666
Chemical Name or Material MM 11253
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.