accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OClNOO

CAS RN 346664-00-4

IUPAC Name: 4-chloro-N,N-diethyl-3-nitrobenzamide
Synonyms: 4-chloro-N,N-diethyl-3-nitrobenzamide
Molecular Weight (g/mol): 256.69
Molecular Formula: C11H13ClN2O3
InChi Key: HWWLUMZDGRNXLU-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results