accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOHNNHNN(E/Z)

CAS RN 346673-06-1

IUPAC Name: 5-(3-hydroxy-4-phenylbut-1-en-1-yl)-1-[6-(2H-1,2,3,4-tetrazol-5-yl)hexyl]pyrrolidin-2-one
Synonyms: 5-(3-hydroxy-4-phenylbut-1-en-1-yl)-1-[6-(2H-1,2,3,4-tetrazol-5-yl)hexyl]pyrrolidin-2-one
Molecular Weight (g/mol): 383.5
Molecular Formula: C21H29N5O2
InChi Key: JWYPJSNXPZTEHL-UHFFFAOYNA-N
SMILES: OC(CC1=CC=CC=C1)C=CC1CCC(=O)N1CCCCCCC1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)