CAS RN 347323-96-0
IUPAC Name:
4-[(2,5-dimethyl-1-phenyl-1H-pyrrol-3-yl)methylidene]-5-thia-2,7-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),6,8,10-tetraen-3-one
Synonyms:
4-[(2,5-dimethyl-1-phenylpyrrol-3-yl)methylidene]-5-thia-2,7-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),6,8,10-tetraen-3-one
Molecular Weight (g/mol):
371.46
Molecular Formula:
C22H17N3OS
InChi Key:
SJMYMKPBODEZSH-UHFFFAOYSA-N
SMILES:
CC1=CC(C=C2SC3=NC4=CC=CC=C4N3C2=O)=C(C)N1C1=CC=CC=C1
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Trio N-Terminal RhoGEF Domain Inhibitor, ITX3
The Trio N-Terminal RhoGEF Domain Inhibitor, ITX3, also referenced under CAS 347323-96-0, controls the biological activity of Trio N-Terminal RhoGEF Domain. This small molecule/inhibitor is primarily used for Cell Structure applications.