accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOBr

CAS RN 34743-88-9

IUPAC Name: 2-(4-bromophenoxy)ethan-1-ol
Synonyms: 2-(4-bromophenoxy)ethanol
Molecular Weight (g/mol): 217.06
Molecular Formula: C8H9BrO2
InChi Key: QYIOGYCRGNHDNK-UHFFFAOYSA-N
SMILES: OCCOC1=CC=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results