accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34770-60-0

IUPAC Name: 4-methylpiperazin-2-one
Synonyms: 4-methylpiperazin-2-one
Molecular Weight (g/mol): 114.15
Molecular Formula: C5H10N2O
InChi Key: KVIZTDNKHOCNAM-UHFFFAOYSA-N
SMILES: CN1CCNC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results