accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3SSiH3CH3CCH3FF

CAS RN 347838-12-4

IUPAC Name: [3,4-difluoro-5-(trimethylsilyl)thiophen-2-yl]trimethylsilane
Synonyms: [3,4-difluoro-5-(trimethylsilyl)thiophen-2-yl]trimethylsilane
Molecular Weight (g/mol): 264.48
Molecular Formula: C10H18F2SSi2
InChi Key: BIIDYHMAKPKADL-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=C(F)C(F)=C(S1)[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results