accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3480-87-3

IUPAC Name: 3-hydroxy-3-phenylpropanoic acid
Synonyms: 3-hydroxy-3-phenylpropanoic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: AYOLELPCNDVZKZ-UHFFFAOYNA-N
SMILES: OC(CC(O)=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results