CAS RN 348086-71-5
IUPAC Name:
N-methyl-N-(4-methyl-5-sulfamoyl-1,3-thiazol-2-yl)-2-[4-(pyridin-2-yl)phenyl]acetamide
Synonyms:
N-methyl-N-(4-methyl-5-sulfamoyl-1,3-thiazol-2-yl)-2-[4-(pyridin-2-yl)phenyl]acetamide
Molecular Weight (g/mol):
402.49
Molecular Formula:
C18H18N4O3S2
InChi Key:
IVZKZONQVYTCKC-UHFFFAOYSA-N
SMILES:
CN(C(=O)CC1=CC=C(C=C1)C1=CC=CC=N1)C1=NC(C)=C(S1)S(N)(=O)=O
1
–
2
of
2
results
1
Promotions Available
Pritelivir, Thermo Scientific™
Pritelivir, Thermo Scientific™
CAS: 348086-71-5 Molecular Formula: C18 H18 N4 O3 S2
2
R&D Systems™ Pritelivir
A Tocris product, part of R&D Systems. Potent and selective herpes simplex virus (HSV) helicase primase inhibitor