accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3486-35-9

IUPAC Name: zinc(2+) carbonate
Synonyms: zinc carbonate
Molecular Weight (g/mol): 125.39
Molecular Formula: CO3Zn
InChi Key: FMRLDPWIRHBCCC-UHFFFAOYSA-L
SMILES: [Zn++].[O-]C([O-])=O
Quantity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)