accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CNHONNONONCH3CH3

CAS RN 349085-38-7

IUPAC Name: 2-(1,3-dimethyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-7-yl)-N-[4-(propan-2-yl)phenyl]acetamide
Synonyms: 2-(1,3-dimethyl-2,6-dioxopurin-7-yl)-N-(4-isopropylphenyl)acetamide
Molecular Weight (g/mol): 355.4
Molecular Formula: C18H21N5O3
InChi Key: HEQDZPHDVAOBLN-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(NC(=O)CN2C=NC3=C2C(=O)N(C)C(=O)N3C)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)