accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONOOCl

CAS RN 349093-97-6

IUPAC Name: N-benzyl-2-chloro-N-ethyl-4-nitrobenzamide
Synonyms: N-benzyl-2-chloro-N-ethyl-4-nitrobenzamide
Molecular Weight (g/mol): 318.76
Molecular Formula: C16H15ClN2O3
InChi Key: XXEMVWJWUFWUQQ-UHFFFAOYSA-N
SMILES: CCN(CC1=CC=CC=C1)C(=O)C1=C(Cl)C=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results