accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CCH3NHOHO

CAS RN 3506-09-0

IUPAC Name: hydrogen 1-[(2-methylpropyl)amino]-3-(naphthalen-1-yloxy)propan-2-ol chloride
Synonyms: hydrogen 1-[(2-methylpropyl)amino]-3-(naphthalen-1-yloxy)propan-2-ol chloride
Molecular Weight (g/mol): 309.83
Molecular Formula: C17H24ClNO2
InChi Key: RTTYPFXTLKDXPS-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)CNCC(O)COC1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results