accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClClCl

CAS RN 35065-28-2

IUPAC Name: 2,2',3,4,4',5'-hexachloro-1,1'-biphenyl
Synonyms: 2,2',3,4,4',5'-hexachloro-1,1'-biphenyl
Molecular Weight (g/mol): 360.86
Molecular Formula: C12H4Cl6
InChi Key: RPUMZMSNLZHIGZ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C(Cl)=C1Cl)C1=CC(Cl)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results