CAS RN 352-24-9
IUPAC Name:
ethyl 4,4-difluoro-3-oxobutanoate
Synonyms:
ethyl 4,4-difluoro-3-oxobutanoate
Molecular Weight (g/mol):
166.12
Molecular Formula:
C6H8F2O3
InChi Key:
CBDPWKVOPADMJC-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC(=O)C(F)F