accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSOHOOCH3(R)OO(R)CH3(S)O(R)(R)H3COH3C(S)OH(S)CH3O(S)CH3(R)O(S)O(R)CH3(S)(R)OOCH3NH3CCH3(R)CH3HO(R)H3CO(R)CH3(R)HO(S)H3COH

CAS RN 3521-62-8

IUPAC Name: (2S,3R,4S,6R)-4-(dimethylamino)-2-{[(3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-2,10-dioxo-1-oxacyclotetradecan-6-yl]oxy}-6-methyloxan-3-yl propanoate; (dodecyloxy)sulfonic acid
Synonyms: ilosone marcoeritrex erythromycin estolate stellamicina eritroger propionylerythromycin lauryl sulfate estomicina PELS lauromicina erythromycin estorate
Molecular Weight (g/mol): 1056.4
Molecular Formula: C52H97NO18S
InChi Key: AWMFUEJKWXESNL-JZBHMOKNSA-N
SMILES: CCCCCCCCCCCCOS(O)(=O)=O.CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2OC(=O)CC)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O
Color 
  • (2)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)