Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythromycin Estolate, G-Biosciences
SCI_ED

Catalog No. S139720
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139720 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139720 Supplier G-Biosciences Supplier No. RC1012
Add to Cart
Add to Cart

Erythromycin Estolate is a macrolide antibiotic isolated from Streptomyces erythreus. It displays strong efficacy against gram-positive cocci and bacilli and some mycobacteria, mycoplasma, and spirochetes.

In addition to its antimicrobial properties, erythromycin is a motilin receptor agonist that has been shown to inhibit respiratory glycoconjugate secretion in vitro.

Chemical Identifiers

CAS 3521-62-8

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 5 g
Percent Purity 98%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.