Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythromycin estolate, 95%, Thermo Scientific™

Catalog No. AAJ6654806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6654806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6654806 Supplier Thermo Scientific Chemicals Supplier No. J6654806

missing translation for 'validMainframeMsg'

A macrolide antibiotic with a target spectrum similar to penicillin

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3521-62-8
Molecular Formula C52H97NO18S
Molecular Weight (g/mol) 1056.40
MDL Number MFCD00084691
InChI Key AWMFUEJKWXESNL-JZBHMOKNSA-N
Synonym Erythromycin 2'-propionate dodecyl sulfate
PubChem CID 133611978
IUPAC Name (2S,3R,4S,6R)-4-(dimethylamino)-2-{[(3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-2,10-dioxo-1-oxacyclotetradecan-6-yl]oxy}-6-methyloxan-3-yl propanoate; (dodecyloxy)sulfonic acid
SMILES CCCCCCCCCCCCOS(O)(=O)=O.CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2OC(=O)CC)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O

Specifications

Type Erythromycin Estolate
Physical Form Powder
Melting Point 135°C to 140°C
Color White
Quantity 5 g
Flash Point >110°C (230°F)
Sensitivity Light sensitive
Boiling Point 743°C
Merck Index 14,3683
Solubility Information Freely soluble in organic solvents,practically insoluble in water /n
Formula Weight 1056.38
Percent Purity 95%
Chemical Name or Material Erythromycin estolate
Show More Show Less
Hazard Category H302-H317-H334
Precautionary Statement P261-P264b-P270-P272-P280g-P285-P301+P312-P302+P352-P304+P341-P330-P333+P313-P342+P311-P363-P501c
EINECSNumber 222-532-4
RTECSNumber KF5775000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.