accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHN

CAS RN 3524-87-6

IUPAC Name: 6-methyl-1,4-dihydropyrimidin-4-one
Synonyms: 4(1H)-pyrimidinone, 6-methyl-
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: LHRIUKSRPHFASO-UHFFFAOYSA-N
SMILES: CC1=CC(=O)N=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results