Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-6-methylpyrimidine, 97%, Thermo Scientific™

Catalog No. AAH6140403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6140403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6140403 Supplier Thermo Scientific Chemicals Supplier No. H6140403

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3524-87-6
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD00053575
InChI Key LHRIUKSRPHFASO-UHFFFAOYSA-N
Synonym 4-hydroxy-6-methylpyrimidine, 6-methylpyrimidin-4-ol, 6-methylpyrimidin-4 3h-one, 4 1h-pyrimidinone, 6-methyl, 6-methyl-4-pyrimidinol, 4-pyrimidinol, 6-methyl, 6-methyl-4 3h-pyrimidinone, 4-pyrimidinol,6-methyl, 6-methyl-4-oxopyrimidine, 4 3h-pyrimidinone, 6-methyl
PubChem CID 19043
IUPAC Name 6-methyl-1H-pyrimidin-4-one
SMILES CC1=CC(=O)N=CN1

Specifications

Melting Point 145°C to 150°C
Color White to Brown
Quantity 1 g
Formula Weight 110.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Hydroxy-6-methylpyrimidine
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
EINECSNumber 222-543-4
RTECSNumber UW7528000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.