accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3529-09-7

IUPAC Name: (2-aminoethyl)dibutylamine
Synonyms: (2-aminoethyl)dibutylamine
Molecular Weight (g/mol): 172.32
Molecular Formula: C10H24N2
InChi Key: PWNDYKKNXVKQJO-UHFFFAOYSA-N
SMILES: CCCCN(CCN)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results