CAS RN 353229-59-1
IUPAC Name:
2,2,2-trifluoro-N-[4-(2-methoxyphenoxy)phenyl]-N-[(pyridin-3-yl)methyl]ethane-1-sulfonamide hydrochloride
Synonyms:
2,2,2-trifluoro-N-[4-(2-methoxyphenoxy)phenyl]-N-(pyridin-3-ylmethyl)ethanesulfonamide hydrochloride
Molecular Weight (g/mol):
488.91
Molecular Formula:
C21H20ClF3N2O4S
InChi Key:
LPWFRDWTOKLHJC-UHFFFAOYSA-N
SMILES:
Cl.COC1=CC=CC=C1OC1=CC=C(C=C1)N(CC1=CN=CC=C1)S(=O)(=O)CC(F)(F)F
1
–
1
of
1
results
1
R&D Systems™ LY 487379 hydrochloride
CAS: 353229-59-1 Molecular Formula: C21H20ClF3N2O4S