accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 353791-85-2

IUPAC Name: 1-[(cyclooct-1-en-1-yl)methyl]-4-(2,7-dichloro-9H-xanthene-9-amido)-1-ethylpiperidin-1-ium iodide
Synonyms: 1-(cyclooct-1-en-1-ylmethyl)-4-(2,7-dichloro-9H-xanthene-9-amido)-1-ethylpiperidin-1-ium iodide
Molecular Weight (g/mol): 655.44
Molecular Formula: C30H37Cl2IN2O2
InChi Key: FOAFBMYSXIGAOX-UHFFFAOYSA-N
SMILES: [I-].CC[N+]1(CC2=CCCCCCC2)CCC(CC1)NC(=O)C1C2=C(OC3=C1C=C(Cl)C=C3)C=CC(Cl)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results