accessibility menu, dialog, popup

UserName

Viewing profile as user:
FClFFCl

CAS RN 354-23-4

IUPAC Name: 1,2-dichloro-1,1,2-trifluoroethane
Synonyms: 1,2-dichloro-1,1,2-trifluoroethane
Molecular Weight (g/mol): 152.93
Molecular Formula: C2HCl2F3
InChi Key: YMRMDGSNYHCUCL-UHFFFAOYNA-N
SMILES: FC(Cl)C(F)(F)Cl
Element 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)