accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFFOOHOFFF

CAS RN 35480-52-5

IUPAC Name: 2,5-bis(2,2,2-trifluoroethoxy)benzoic acid
Synonyms: 2,5-bis(2,2,2-trifluoroethoxy)benzoic acid
Molecular Weight (g/mol): 318.17
Molecular Formula: C11H8F6O4
InChi Key: YPGYLCZBZKRYQJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(OCC(F)(F)F)C=CC(OCC(F)(F)F)=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)