accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFFFFFF

CAS RN 355-04-4

IUPAC Name: 1,1,1,2,2,3,3,4,5,5,5-undecafluoro-4-(trifluoromethyl)pentane
Synonyms: perfluoroisohexane
Molecular Weight (g/mol): 338.04
Molecular Formula: C6F14
InChi Key: ROVMKEZVKFJNBD-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results