accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFH2N

CAS RN 355-27-1

IUPAC Name: 2,2,3,3,4,4,5,5,5-nonafluoropentan-1-amine
Synonyms: 2,2,3,3,4,4,5,5,5-nonafluoropentan-1-amine
Molecular Weight (g/mol): 249.08
Molecular Formula: C5H4F9N
InChi Key: SUNUNYBGDFIAME-UHFFFAOYSA-N
SMILES: NCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results